Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037916.3 | NP_001033005.1 | 2 | 256 | ||
GenBank mRNA | AK029257.1 | - | 2 | 258 | ||
AK076542.1 | - | 2 | 333 |
Gene Symbol: C530005A16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035052.1 | - | 1 | 572 | ||
AK039789.1 | - | 1 | 570 | |||
AK086654.1 | - | 1 | 569 | |||
AK140096.1 | - | 1 | 548 | |||
AK164083.1 | - | 1 | 532 |