Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt61b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI265689.1 | - | 3 | 528 | ||
AK004173.1 | - | 1 | 16 | |||
AK032413.1 | - | 1 | 509 | |||
AK036113.1 | - | 1 | 518 | |||
AK045842.1 | - | 1 | 470 |