Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022427.2 | NP_071872.1 | 2 | 644 | ||
GenBank mRNA | AB042408.1 | - | 2 | 614 | ||
BC029939.1 | - | 2 | 555 | |||
BC051941.1 | - | 1 | 260 | 5708059 | ||
BC157981.1 | - | 1 | 253 | 9007300 |
Set Membership: |
G-protein coupled receptor |