Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025474.3 | NP_079750.1 | 3 | 325 | ||
GenBank mRNA | AK007673.1 | - | 3 | 338 | ||
AK010242.1 | - | 3 | 331 | |||
AK083352.1 | - | 3 | 325 | |||
AK144195.1 | - | 332 | ||||
BC044057.1 | - | 3 | 319 | 6489566 |