Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtdap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033131.3 | NP_001028303.1 | 2, 3 | 153 | ||
GenBank mRNA | AK131603.1 | - | 2, 3 | 154 | ||
AK131606.1 | - | 2, 3 | 154 | |||
BC132510.1 | - | 2, 3 | 153 | 40130787 | ||
BC132512.1 | - | 2, 3 | 153 | 40130789 | ||
BC145438.1 | - | 2, 3 | 153 | 9053996 |