Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dear1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040461.2 | NP_001035551.2 | 1 | 354 | ||
GenBank mRNA | BC141214.1 | - | 1 | 346 | 9055826 | |
DQ009865.1 | - | 1 | 352 |