Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kncn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039124.3 | NP_001034213.1 | 1 | 423 | ||
GenBank mRNA | BC132034.1 | - | 1 | 370 | 40130311 | |
BC132036.1 | - | 1 | 370 | 40130313 | ||
BC145375.1 | - | 1 | 370 | 9053933 | ||
DQ235452.1 | - | 1 | 423 |