Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025313.2 | NP_079589.2 | 2, 3 | 435 | ||
GenBank mRNA | AK002330.1 | - | 2, 3 | 385 | ||
AK005082.1 | - | 2, 3 | 379 | |||
AK011374.1 | - | 2, 3 | 356 | |||
AK018011.1 | - | 2, 3 | 381 | |||
AK076064.1 | - | 2, 3 | 360 | |||
AK151151.1 | - | 2, 3 | 370 | |||
AK155285.1 | - | 2, 3 | 419 | |||
AK165932.1 | - | 2, 3 | 384 | |||
BC008273.1 | - | 2, 3 | 344 | 2811419 | ||
CT010177.1 | - | 2, 3 | 284 |