Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025317.2 | NP_079593.1 | 2, 3 | 315 | ||
GenBank mRNA | AK002348.1 | - | 2, 3 | 317 | ||
AK003225.1 | - | 2, 3 | 316 | |||
AK009657.1 | - | 2, 3 | 298 | |||
BC056985.1 | - | 2, 3 | 298 | 6818054 |