Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtcap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025327.2 | NP_079603.1 | 5 | 467 | ||
GenBank mRNA | AK002479.1 | - | 5 | 468 | ||
AK003310.1 | - | 5 | 435 | |||
BC031492.1 | - | 5 | 420 | 3485092 | ||
BC043030.1 | - | 5 | 939 | 5693076 |