Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025343.5 | NP_079619.4 | 4, 5 | 812 | ||
GenBank mRNA | AK007323.1 | - | 4, 5 | 760 | ||
AK075603.1 | - | 4, 5 | 795 | |||
AK163523.1 | - | 48 | ||||
BC019572.1 | - | 1, 2 | 57 | |||
BC027299.1 | - | 4, 5 | 760 | 3987394 | ||
BC036150.1 | - | 3, 4 | 732 | 4977394 |