Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025347.2 | NP_079623.1 | 5 | 516 | ||
NM_026875.2 | NP_081151.2 | 5 | 616 | |||
GenBank mRNA | AB098742.1 | - | 4 | 321 | ||
AF523353.1 | - | 5 | 506 | |||
AK002925.1 | - | 5 | 501 | |||
AK003371.1 | - | 5 | 517 | |||
AK004431.1 | - | 5 | 589 | |||
BC009171.1 | - | 5 | 490 |