Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025366.3 | NP_079642.1 | 2 | 215 | ||
GenBank mRNA | AK003263.1 | - | 2 | 174 | ||
AK010311.1 | - | 2 | 158 | |||
AK011261.1 | - | 2 | 173 | |||
AK027907.1 | - | 2 | 199 | |||
AV460497.1 | - | 2 | 215 | |||
BC049582.1 | - | 198 | 6772118 | |||
BC052720.1 | - | 2 | 194 | 6818266 | ||
BC064310.1 | - | 2 | 194 | |||
BC092216.1 | - | 2 | 182 | 30609530 |