Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025401.3 | NP_079677.1 | 3 | 1707 | ||
GenBank mRNA | AK002730.1 | - | 3 | 216 | ||
AK003992.1 | - | 3 | 223 | |||
AK007726.1 | - | 3 | 188 | |||
AK009854.1 | - | 3 | 223 | |||
AK012803.1 | - | 3 | 205 | |||
AK051149.1 | - | 3 | 1708 | |||
AK154036.1 | - | 2 | 2299 | |||
BC028498.1 | - | 3 | 177 | 1400865 |