Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025417.2 | NP_079693.1 | 5 | 265 | ||
GenBank mRNA | AK004174.1 | - | 5 | 266 | ||
AK009802.1 | - | 5 | 264 | |||
AK172194.1 | - | 4 | 1377 | |||
BC049222.1 | - | 5 | 264 | 2649359 | ||
BC055798.1 | - | 4 | 1356 |