Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc28b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025455.2 | NP_079731.2 | 3 | 372 | ||
GenBank mRNA | AK003287.1 | - | 3 | 342 | ||
AK007427.1 | - | 2 | 149 | |||
AK028262.1 | - | 3 | 350 | |||
AK031278.1 | - | 1 | 98 | |||
AK158016.1 | - | 3 | 376 | |||
BB848581.1 | - | 3 | 413 | |||
BC028873.1 | - | 3 | 364 | 4220355 | ||
BQ031123.1 | - | 4 | 481 |