Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tma16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025465.2 | NP_079741.1 | 4 | 262 | ||
GenBank mRNA | AK007637.1 | - | 4 | 262 | ||
AK009167.1 | - | 4 | 277 | |||
AK011532.1 | - | 4 | 273 | |||
BC016246.1 | - | 4 | 308 | 4511374 | ||
BC034844.1 | - | 4 | 258 | 1496036 |