Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025478.3 | NP_079754.2 | 1, 2 | 318 | ||
GenBank mRNA | AK011674.1 | - | 1, 2 | 185 | ||
AK134794.1 | - | 1, 2 | 318 | |||
AK152763.1 | - | 1, 2 | 315 | |||
AK153257.1 | - | 1, 2 | 315 | |||
AK165542.1 | - | 1, 2 | 337 | |||
AK185662.1 | - | 16 | ||||
AY035213.1 | - | 1, 2 | 318 | |||
BC016576.1 | - | 1, 2 | 325 | 2609919 |