Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025512.2 | NP_079788.2 | 5 | 335 | ||
GenBank mRNA | AK009240.1 | - | 5 | 349 | ||
AK017696.1 | - | 5 | 349 | |||
AK030472.1 | - | 5 | 343 | |||
AK077873.1 | - | 5 | 345 | |||
BC023352.1 | - | 5 | 327 | |||
BC025216.1 | - | 5 | 327 | |||
BC046585.1 | - | 5 | 335 | 6314252 | ||
BC089494.1 | - | 5 | 347 | 30504018 |