Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023202.4 | NP_075691.1 | 1 | 72 | ||
GenBank mRNA | AK002476.1 | - | 1 | 53 | ||
AK003468.1 | - | 1 | 53 | |||
AK010299.1 | - | 1 | 30 | |||
BC044054.1 | - | 39 | ||||
BC052817.1 | - | 1 | 36 | 6487401 | ||
BC055698.1 | - | 1 | 37 | 6411262 | ||
BY605952.1 | - | 1 | 29 | |||
KJ650204.1 | - | 1 | 14 |