Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023202.4 | NP_075691.1 | 1, 2 | 116 | ||
GenBank mRNA | AK002476.1 | - | 1, 2 | 97 | ||
AK003468.1 | - | 1, 2 | 97 | |||
AK010299.1 | - | 1, 2 | 74 | |||
BC044054.1 | - | 83 | ||||
BC052817.1 | - | 1, 2 | 80 | 6487401 | ||
BC055698.1 | - | 1, 2 | 81 | 6411262 | ||
BY605952.1 | - | 1, 2 | 73 | |||
KJ650204.1 | - | 1, 2 | 58 |