Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppdpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025598.2 | NP_079874.1 | 5 | 545 | ||
GenBank mRNA | AK002602.1 | - | 4 | 665 | ||
AK003282.1 | - | 5 | 546 | |||
AK012037.1 | - | 5 | 429 | |||
AK012327.1 | - | 5 | 547 | |||
AK012545.1 | - | 3 | 520 | |||
AK014211.1 | - | 4 | 499 | |||
AK154758.1 | - | 5 | 586 | |||
AK166314.1 | - | 509 | ||||
BC029225.1 | - | 4 | 481 | |||
BC058516.1 | - | 5 | 583 | 6824261 | ||
BC091753.1 | - | 5 | 352 | 4218108 | ||
BC100439.1 | - | 5 | 437 | 6531448 |