Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isoc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101598.1 | NP_001095068.1 | 3 | 395 | ||
GenBank mRNA | BC158139.1 | - | 3 | 295 | 9007402 | |
BG404719.1 | - | 3 | 394 |