Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamtsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285435.1 | NP_001272364.1 | 6 | 543 | ||
GenBank mRNA | AK008355.1 | - | 6 | 508 | ||
AK008364.1 | - | 6 | 508 | |||
AK008440.1 | - | 6 | 507 | |||
AK028819.1 | - | 6 | 1015 | |||
AK029081.1 | - | 6 | 484 | |||
AK154947.1 | - | 3 | 133 | |||
AK219627.1 | - | 1 | 39 | |||
BC034843.1 | - | 4 | 396 | |||
BC058577.1 | - | 2 | 1119 | |||
CK624091.1 | - | 1 | 48 |