Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamtsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285435.1 | NP_001272364.1 | 7 | 649 | ||
GenBank mRNA | AK008355.1 | - | 7 | 614 | ||
AK008364.1 | - | 7 | 614 | |||
AK008440.1 | - | 7 | 613 | |||
AK028819.1 | - | 7 | 1121 | |||
AK029081.1 | - | 7 | 590 | |||
AK154947.1 | - | 4 | 239 | |||
BC034843.1 | - | 5 | 502 | |||
BC058577.1 | - | 3 | 1225 | |||
CK624091.1 | - | 2 | 154 |