Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025645.2 | NP_079921.2 | 3 | 416 | ||
GenBank mRNA | AK002371.1 | - | 3 | 380 | ||
AK004277.1 | - | 3 | 346 | |||
AK136135.1 | - | 3 | 346 | |||
AK148443.1 | - | 3 | 355 | |||
AK152287.1 | - | 3 | 380 | |||
AK152352.1 | - | 3 | 374 | |||
AK169605.1 | - | 3 | 362 | |||
BC058365.1 | - | 3 | 348 | 5714866 |