Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogfod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025671.2 | NP_079947.1 | 5 | 417 | ||
GenBank mRNA | AK004916.1 | - | 3 | 885 | ||
AK004943.1 | - | 5 | 418 | |||
AK017497.1 | - | 5 | 418 | |||
AK018534.1 | - | 5 | 418 | |||
AK168946.1 | - | 5 | 426 | |||
BC022762.1 | - | 4 | 265 | |||
BC069872.1 | - | 5 | 420 | 4219293 |