Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H60b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177775.1 | NP_001171246.1 | 5 | 691 | ||
GenBank mRNA | AB284505.1 | - | 768 |