Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syce1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048145.1 | NP_001041610.1 | 2 | 108 | ||
NM_029139.1 | NP_083415.1 | 2 | 108 | |||
GenBank mRNA | AK015606.1 | - | 2 | 104 | ||
AY899935.1 | - | 2 | 108 | |||
AY899936.1 | - | 2 | 108 | |||
BB616021.1 | - | 2 | 88 | |||
BC141326.1 | - | 2 | 108 | 9055938 | ||
BC141347.1 | - | 2 | 108 | 9055959 |