Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033851.1 | NP_001029023.1 | 1 | 125 | ||
NM_025815.2 | NP_080091.1 | 1 | 125 | |||
GenBank mRNA | AK004559.1 | - | 1 | 113 | ||
AK005311.1 | - | 1 | 59 | |||
AK010227.1 | - | 1 | 125 | |||
AK013407.1 | - | 1 | 110 | |||
BC017626.1 | - | 1 | 96 | 4502073 | ||
BC048551.1 | - | 1 | 125 | |||
BC076564.1 | - | 1 | 84 | |||
BC138745.1 | - | 1 | 124 | 8861767 | ||
BC138746.1 | - | 1 | 124 | 8861768 | ||
BU846828.1 | - | 129 |