Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025910.3 | NP_080186.3 | 8 | 1275 | ||
GenBank mRNA | AB177385.1 | - | 7 | 1076 | ||
AK007810.1 | - | 8 | 1245 | |||
AK013451.1 | - | 8 | 1235 | |||
AK031671.1 | - | 8 | 1209 | |||
BC023462.1 | - | 8 | 1530 | 5041558 | ||
BC025109.1 | - | 8 | 1197 | 4236994 | ||
BC058242.1 | - | 8 | 1184 | 6308890 |