Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025920.3 | NP_080196.3 | 6 | 1973 | ||
GenBank mRNA | AK008304.1 | - | 4 | 357 | ||
AK008587.1 | - | 4 | 356 | |||
AK140888.1 | - | 1916 | ||||
AK167814.1 | - | 6 | 1733 | |||
BC013538.1 | - | 5 | 810 | |||
BC057963.1 | - | 5 | 1271 | |||
BC063758.1 | - | 6 | 1704 | 6334411 | ||
BC066042.1 | - | 6 | 1908 | 6413779 |