Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277273.1 | NP_001264202.1 | 5 | 451 | ||
NM_025923.3 | NP_080199.1 | 5 | 451 | |||
GenBank mRNA | AA476004.1 | - | 3 | 122 | ||
AF513619.1 | - | 5 | 451 | |||
AK008605.1 | - | 5 | 337 | |||
AK011520.1 | - | 5 | 356 | |||
AK011752.1 | - | 5 | 328 | |||
AK011903.1 | - | 5 | 334 | |||
AK017431.1 | - | 5 | 334 | |||
AK018018.1 | - | 5 | 333 | |||
AK076017.1 | - | 5 | 334 | |||
AK168538.1 | - | 5 | 335 | |||
BC032876.1 | - | 5 | 325 | 1282316 | ||
CN455644.1 | - | 4 | 237 |