Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027349.3 | NP_081625.3 | 7 | 805 | ||
GenBank mRNA | AK011403.1 | - | 7 | 786 | ||
AK142132.1 | - | 1 | 81 | |||
AK151910.1 | - | 7 | 757 | |||
AK162135.1 | - | 898 | ||||
AK164367.1 | - | 7 | 770 | |||
AK169022.1 | - | 1 | 962 | |||
BC010792.1 | - | 6 | 551 | |||
BC023749.1 | - | 5 | 514 | |||
BC158104.1 | - | 6 | 685 | 9007388 |