Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Higd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025933.3 | NP_080209.1 | 2 | 304 | ||
GenBank mRNA | AK004272.1 | - | 2 | 144 | ||
AK005269.1 | - | 2 | 305 | |||
AK008387.1 | - | 2 | 301 | |||
BC021471.1 | - | 2 | 301 | 5343356 |