Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025960.3 | NP_080236.2 | 3 | 265 | ||
GenBank mRNA | AK007965.1 | - | 3 | 264 | ||
AK008022.1 | - | 3 | 265 | |||
AK008462.2 | - | 3 | 264 | |||
AK088832.1 | - | 3 | 264 | |||
BC020128.1 | - | 2 | 214 | |||
BC037154.1 | - | 3 | 302 | 2811950 | ||
BC106114.1 | - | 3 | 279 | 4168174 |