Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026014.3 | NP_080290.3 | 5 | 784 | ||
GenBank mRNA | AB086655.1 | - | 5 | 763 | ||
AF477481.1 | - | 5 | 784 | |||
AK012041.1 | - | 5 | 761 | |||
AK017609.1 | - | 5 | 758 | |||
AK028287.1 | - | 5 | 756 | |||
AK049163.1 | - | 5 | 756 | |||
AK076527.1 | - | 4 | 866 | |||
AK083015.1 | - | 5 | 759 | |||
AK198035.1 | - | 1 | 205 | |||
AK204196.1 | - | 1 | 205 | |||
BC013842.1 | - | 5 | 739 | |||
BC024485.1 | - | 2 | 35 | |||
BC048076.1 | - | 5 | 784 | 6307476 |