Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023322.2 | NP_075811.2 | 1 | 245 | ||
GenBank mRNA | AK009842.1 | - | 1 | 232 | ||
AK013258.1 | - | 1 | 233 | |||
AK138775.1 | - | 1 | 232 | |||
AK153972.1 | - | 1 | 228 | |||
BC027512.1 | - | 1 | 209 | |||
BC099448.1 | - | 1 | 163 | |||
BC141244.1 | - | 1 | 171 | 9055856 | ||
CJ072232.1 | - | 1 | 245 | |||
U62906.1 | - | 1 | 218 |