Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026069.3 | NP_080345.1 | 3, 4 | 301 | ||
GenBank mRNA | AK002592.1 | - | 3, 4 | 270 | ||
AK008563.1 | - | 3, 4 | 270 | |||
AK011101.1 | - | 3, 4 | 262 | |||
AK012544.1 | - | 3, 4 | 270 | |||
AK014003.1 | - | 3, 4 | 270 | |||
AK019020.1 | - | 3, 4 | 270 | |||
AK019294.1 | - | 3, 4 | 268 | |||
AK028138.1 | - | 3, 4 | 260 | |||
AK168550.1 | - | 3, 4 | 270 | |||
BC034524.1 | - | 248 | ||||
BC054388.1 | - | 3, 4 | 257 | 6478626 | ||
BC081438.1 | - | 2, 3 | 232 | 6817584 |
Gene Symbol: Rpl37rt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271590.1 | NP_001258519.1 | 2 | 315 | ||
NM_001271591.1 | NP_001258520.1 | 2 | 309 | |||
GenBank mRNA | BB707379.1 | - | 2 | 311 | ||
CO799256.1 | - | 347 | ||||
CO814488.2 | - | 1 | 201 |