Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqcf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026090.2 | NP_080366.2 | 3 | 683 | ||
GenBank mRNA | AK006688.1 | - | 3 | 682 | ||
BC048652.1 | - | 3 | 689 | 6772797 | ||
BC061014.1 | - | 3 | 683 | 6770556 | ||
DQ153246.1 | - | 3 | 712 |