Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026095.4 | NP_080371.1 | 2 | 158 | ||
GenBank mRNA | AK005341.1 | - | 2 | 180 | ||
AK006692.1 | - | 2 | 158 | |||
AK160321.1 | - | 2 | 652 | |||
BC011510.1 | - | 2 | 104 | 3256792 |