Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paxbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026110.2 | NP_080386.3 | 8 | 1493 | ||
GenBank mRNA | AK007365.1 | - | 2 | 181 | ||
AK079394.1 | - | 1 | 340 | |||
AY033907.1 | - | 8 | 1278 | |||
AY033908.1 | - | 8 | 1278 |