Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168659.1 | NP_001162130.1 | 3 | 804 | ||
NM_024201.3 | NP_077163.1 | 3 | 798 | |||
GenBank mRNA | AK002564.1 | - | 3 | 786 | ||
AK007738.1 | - | 3 | 750 | |||
AK170932.1 | - | 3 | 791 | |||
BC003345.1 | - | 3 | 778 | 3257525 | ||
BC049099.1 | - | 3 | 756 | 5721356 |