Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026156.2 | NP_080432.1 | 14 | 1901 | ||
GenBank mRNA | AK002890.1 | - | 14 | 1902 | ||
AK031533.1 | - | 1988 | ||||
AK087952.1 | - | 11 | 2164 | |||
AK132434.1 | - | 14 | 1915 | |||
AK145925.1 | - | 14 | 1903 | |||
BC021341.1 | - | 14 | 1899 | 5066146 |