Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026191.2 | NP_080467.3 | 11 | 1516 | ||
GenBank mRNA | AK004610.1 | - | 11 | 1480 | ||
AK010512.1 | - | 11 | 1496 | |||
AK029935.1 | - | 11 | 1487 | |||
AK031000.1 | - | 11 | 1462 | |||
AK075991.1 | - | 5 | 522 | |||
AK135592.1 | - | 11 | 1453 | |||
AK208938.1 | - | 1 | 30 | |||
BC058276.1 | - | 11 | 1434 | 6515479 |