Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026232.3 | NP_080508.1 | 8 | 854 | ||
GenBank mRNA | AK015679.1 | - | 8 | 855 | ||
AK017035.1 | - | 8 | 855 | |||
AK030950.1 | - | 8 | 823 | |||
AK090086.1 | - | 8 | 791 | |||
AK151334.1 | - | 8 | 823 | |||
AK151420.1 | - | 8 | 791 | |||
AK159122.1 | - | 8 | 852 | |||
AK159732.1 | - | 8 | 852 |