Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyrm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163628.1 | NP_001157100.1 | 2 | 414 | ||
NM_133688.3 | NP_598449.1 | 2 | 177 | |||
GenBank mRNA | AF412298.1 | - | 2 | 831 | ||
AF412299.1 | - | 3 | 412 | |||
AF412300.1 | - | 2 | 178 | |||
AK015530.1 | - | 2 | 150 | |||
AK075835.1 | - | 2 | 158 | |||
AY753325.1 | - | 2 | 169 | |||
BC021522.1 | - | 2 | 125 | 5352723 | ||
CX203242.1 | - | 2 | 414 |