Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026591.3 | NP_080867.2 | 2 | 284 | ||
GenBank mRNA | AK003362.1 | - | 2 | 288 | ||
AK008117.1 | - | 2 | 254 | |||
AK013394.1 | - | 2 | 228 | |||
AK016190.1 | - | 2 | 283 | |||
AK019976.1 | - | 2 | 285 | |||
AK020157.1 | - | 2 | 285 | |||
AK167623.1 | - | 2 | 446 | |||
BC004736.1 | - | 1 | 198 | 3498625 | ||
BC025506.1 | - | 2 | 297 | 5251655 | ||
BC031730.1 | - | 2 | 211 | 3991689 |