Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001026211.2 | NP_001021382.1 | 5 | 417 | ||
NM_001286972.1 | NP_001273901.1 | 5 | 417 | |||
NM_001286973.1 | NP_001273902.1 | 4 | 365 | |||
NM_026332.4 | NP_080608.3 | 5 | 417 | |||
GenBank mRNA | AK007866.1 | - | 5 | 385 | ||
AK008267.1 | - | 5 | 361 | |||
AK010204.1 | - | 5 | 367 | |||
AK012051.1 | - | 5 | 369 | |||
AK076602.1 | - | 376 | ||||
BC020327.1 | - | 5 | 350 | |||
BC024335.1 | - | 5 | 325 | 4974988 | ||
CA469052.1 | - | 5 | 454 | |||
CJ123658.1 | - | 4 | 322 | |||
EL962106.1 | - | 5 | 363 |