Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164725.1 | NP_001158197.1 | 4 | 670 | ||
GenBank mRNA | DQ470838.1 | - | 4 | 675 | ||
DQ470839.1 | - | 4 | 675 | |||
DQ470840.1 | - | 4 | 666 | |||
EF032497.1 | - | 4 | 579 |